Fmoc-N-Me-Phe-OH 98,0 HPLC, T

Supplier: TCI America

Synonyms: N-Fmoc-N-methyl-L-phenylalanine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-phenylpropanoic acid, Fmoc-N-Me-Phe-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-phenylalanine

F1167-1G F1167-5G
TCF1167-1GEA 64.78 CAD
TCF1167-1G TCF1167-5G
Fmoc-N-Me-Phe-OH 98,0 HPLC, T
Fmoc-N-Me-Phe-OH
Formula: C₂₅H₂₃NO₄
MW: 401.462 g/mol
MDL Number: MFCD00151938
CAS Number: 77128-73-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 132.0 to 136.0 deg-C
Specific rotation [a]20/D -58.0 to -62.0 deg (C=1,DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR