Fmoc-Sar-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F1163-25G F1163-5G
TCF1163-25GEA 273.34 CAD
TCF1163-25G TCF1163-5G
Fmoc-Sar-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-Sar-OH
Formula: C₁₈H₁₇NO₄
MDL Number: MFCD00151926
CAS Number: 77128-70-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR