N-Fmoc-N'-trityl-D-histidine ≥97.0% (by HPLC)
Supplier: TCI America
F1145-1G
F1145-5G
TCF1145-1GEA
127.98
CAD
TCF1145-1G
TCF1145-5G
N-Fmoc-N'-trityl-D-histidine ≥97.0% (by HPLC)
N-Fmoc-N'-trityl-D-histidine
Formula:
C₄₀H₃₃N₃O₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00077061 CAS Number: 135610-90-1 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Specific rotation [a]20/D | -85.0 to -95.0 deg (C=5, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...