Fmoc-Lys(Fmoc)-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Nalpha,Nepsilon-Bis[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine, Fmoc-Lys(Fmoc)-OH, Nalpha,Nepsilon-Di-Fmoc-L-lysine

B4931-25G B4931-5G
TCB4931-25GEA 239.56 CAD
TCB4931-25G TCB4931-5G
Fmoc-Lys(Fmoc)-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-Lys(Fmoc)-OH
Formula: C₃₆H₃₄N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00080275
CAS Number: 78081-87-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 133.0 to 137.0 deg-C
Specific rotation [a]20/D -6.0 to -8.0 deg (C=1,DMF)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR