Boc-N-Me-D-Phe-OH 98,0 HPLC, T

Supplier: TCI America

Synonyms: N-Boc-N-methyl-D-phenylalanine, N-(tert-Butoxycarbonyl)-N-methyl-D-phenylalanine, Boc-N-Me-D-Phe-OH

B5011-1G B5011-5G
TCB5011-1GEA 93.22 CAD
TCB5011-1G TCB5011-5G
Boc-N-Me-D-Phe-OH 98,0 HPLC, T
Boc-N-Me-D-Phe-OH
Formula: C₁₅H₂₁NO₄
MW: 279.336 g/mol
MDL Number: MFCD00151890
CAS Number: 85466-66-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 84.0 to 88.0 deg-C
Specific rotation [a]20/D +85.0 to +89.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR