Boc-D-Tyr(ME)-OH 98,0 Titrimetric

Supplier: TCI America

Synonyms: Boc-D-Phe(4-OMe)-OH, N-Boc-4-methoxy-D-phenylalanine, N-(tert-Butoxycarbonyl)-4-methoxy-D-phenylalanine

B5350-1G B5350-5G
TCB5350-1GEA 94.8 CAD
TCB5350-1G TCB5350-5G
Boc-D-Tyr(ME)-OH 98,0 Titrimetric
Boc-D-Tyr(ME)-OH
Formula: C₁₅H₂₁NO₅
MW: 295.335 g/mol
MDL Number: MFCD00065604
CAS Number: 68856-96-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 93.0 to 97.0 deg-C
Specific rotation [a]20/D -30.0 to -34.0 deg (C=1, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR