Boc-Asp-OtBu 95,0 HPLC, T

Supplier: TCI America

Synonyms: Boc-Asp-OtBu, N-Boc-L-aspartic acid 1-tert-Butyl ester, N-(tert-Butoxycarbonyl)-L-aspartic acid 1-tert-Butyl ester, 1-tert-Butyl N-Boc-L-aspartate, 1-tert-Butyl N-(tert-Butoxycarbonyl)-L-aspartate, N-Boc-L-aspartic acid 1-tert-butyl ester

B4618-1G B4618-5G
TCB4618-1GEA 74.26 CAD
TCB4618-1G TCB4618-5G
Boc-Asp-OtBu 95,0 HPLC, T
Boc-Asp-OtBu
Formula: C₁₃H₂₃NO₆
MW: 289.328 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038272
CAS Number: 34582-32-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 % (Exclude toluene)
Purity (Neutralization titration) min. 95.0 %
Melting point 101.0 to 106.0 deg-C
Specific rotation [a]20/D -22.0 to -25.0 deg (C=1.5, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR