Boc-Tic-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

B4501-25G B4501-5G
76219-394EA 395 CAD
76219-394 76219-396
Boc-Tic-OH ≥98.0% (by HPLC, titration analysis)
Boc-Tic-OH
Formula: C₁₅H₁₉NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00143845
CAS Number: 78879-20-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 129.0 to 133.0 deg-C
Specific rotation [a]20/D +19.0 to +22.0 deg (C=5,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR