Polyethylene glycol monomethyl ether 1000
Supplier: TCI America
P2185-100G
P2185-500G
TCP2185-100GEA
42.66
CAD
TCP2185-100G
TCP2185-500G
Polyethylene glycol monomethyl ether 1000
Polyethylene glycol monomethyl ether
Formula:
CH₃(OCH₂CH₂)nOH |
MDL Number:
MFCD00084416 CAS Number: 9004-74-4 |
Specification Test Results
Appearance | White to Almost white powder to lump |
Average molecular weight | 950 to 1,050 |
Hydroxyl Value | 50.0 to 60.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...