Polyethylene glycol monomethyl ether 400

Supplier: TCI America

Synonyms: Methoxypolyethylene glycols, Methoxy poly(ethylene glycol), PEG MME, Poly(ethylene glycol) methyl ether, mPEG, Methoxypolyethylene glycols-average M.W. 1000, m-PEG-OH

P2183-100G P2183-500G
TCP2183-100GEA 41.81 CAD
TCP2183-100G TCP2183-500G
Polyethylene glycol monomethyl ether 400
Polyethylene glycol monomethyl ether
Formula: CH₃(OCH₂CH₂)nOH
MDL Number: MFCD00084416
CAS Number: 9004-74-4

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Average molecular weight 380 to 420
Hydroxyl Value 130 to 150


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR