Piperazine dihydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Danger

P2491-1G P2491-5G
TCP2491-1GEA 105.86 CAD
TCP2491-1G TCP2491-5G
Piperazine dihydrochloride ≥98.0% (by total nitrogen and titration analysis)
Piperazine dihydrochloride
Formula: C₄H₁₀N₂·2HCl
Melting Pt: 320°C
MDL Number: MFCD00012754
CAS Number: 142-64-3
EINECS: 205-551-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR