N,N,N',N'-Tetraphenylbenzidine ≥99.0% (by GC), purified by sublimation

Supplier: TCI America

Synonyms: TPB, N,N,N,N-Tetraphenylbenzidine, 4,4'-Bis(diphenylamino)biphenyl

T3266-1G T3266-5G
TCT3266-1GEA 126.4 CAD
TCT3266-1G TCT3266-5G
N,N,N',N'-Tetraphenylbenzidine ≥99.0% (by GC), purified by sublimation
N,N,N',N'-Tetraphenylbenzidine
Formula: C₃₆H₂₈N₂
MW: 488.63 g/mol
MDL Number: MFCD00228123
CAS Number: 15546-43-7
EINECS: 239-599-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 231.0 to 236.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR