Dansylglycine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D5406-100MG D5406-25MG
TCD5406-100MGEA 64.78 CAD
TCD5406-100MG TCD5406-25MG
Dansylglycine ≥98.0% (by titrimetric analysis)
Dansylglycine
Formula: C₁₄H₁₆N₂O₄S
MDL Number: MFCD00037734
CAS Number: 1091-85-6
EINECS: 214-129-7

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 156.0 to 161.0 deg-C
Human serum albmin (HSA) binding activity S/N ratio min. 8


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR