1H,1H,5H-Octafluoropentyl methacrylate ≥98.0% (by GC) stabilised

Supplier: TCI America
Warning

Synonyms: Methacrylic Acid 1H,1H,5H-Perfluoropentyl Ester, Methacrylic Acid 1H,1H,5H-Octafluoropentyl Ester, 1H,1H,5H-Perfluoropentyl Methacrylate, 2,2,3,3,4,4,5,5-Octafluoropentyl methacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

O0481-25G O0481-5G
76220-188EA 189.6 CAD
76220-188 76220-190
1H,1H,5H-Octafluoropentyl methacrylate ≥98.0% (by GC) stabilised
1H,1H,5H-Octafluoropentyl methacrylate
Formula: C₉H₈F₈O₂
Boiling Pt: 88°C
Flash Pt: 81°C
MDL Number: MFCD00039278
CAS Number: 355-93-1
EINECS: 206-596-0

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR