1,2,4,5-Cyclohexanetetracarboxylic dianhydride ≥98.0% (by titrimetric analysis), purified by sublimation

Supplier: TCI America
Warning

C2919-1G C2919-5G
76218-890EA 145.36 CAD
76218-890 76218-892
1,2,4,5-Cyclohexanetetracarboxylic dianhydride ≥98.0% (by titrimetric analysis), purified by sublimation
1,2,4,5-Cyclohexanetetracarboxylic dianhydride
Formula: C₁₀H₈O₆
Melting Pt: 240°C
MDL Number: MFCD13704912
CAS Number: 2754-41-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Anilin Method) min. 98.0 %
Solubility in Acetone almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR