1,2,3,4-Cyclopentanetetracarboxylic dianhydride ≥99.0% (by titrimetric analysis), purified by sublimation

Supplier: TCI America
Warning

C2920-1G C2920-5G
TCC2920-1GEA 115.66 CAD
TCC2920-1G TCC2920-5G
1,2,3,4-Cyclopentanetetracarboxylic dianhydride ≥99.0% (by titrimetric analysis), purified by sublimation
1,2,3,4-Cyclopentanetetracarboxylic dianhydride
Formula: C₉H₆O₆
MW: 210.14 g/mol
MDL Number: MFCD00046872
CAS Number: 6053-68-5
EINECS: 227-964-7

Order Now

Specification Test Results

Appearance White powder to crystal
Purity (Neutralization titration) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR