Ammonium ((1S,3R,4R,7S)-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl)methanesulfonate ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B4412-1G B4412-5G
TCB4412-1GEA 104.28 CAD
TCB4412-1G TCB4412-5G
Ammonium ((1S,3R,4R,7S)-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl)methanesulfonate ≥97.0% (by titrimetric analysis)
Ammonium ((1S,3R,4R,7S)-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl)methanesulfonate
Formula: C₁₀H₁₈BrNO₄S
Storage Temperature: Freezer
MDL Number: MFCD00151070
CAS Number: 55870-50-3
EINECS: 259-871-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (Ion exchange titration) min. 97.0 %
Specific rotation [a]20/D -83.0 to -86.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR