Ethyl-1,3-benzothiazole-2-carboxylate ≥98.0% (by GC)
Supplier: TCI America
E1089-25G
E1089-5G
TCE1089-25GEA
246.32
CAD
TCE1089-25G
TCE1089-5G
Ethyl-1,3-benzothiazole-2-carboxylate ≥98.0% (by GC)
Ethyl-1,3-benzothiazole-2-carboxylate
Formula:
C₁₀H₉NO₂S |
MDL Number:
MFCD00848360 CAS Number: 32137-76-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 68.0 to 72.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...