tert-Butyl-6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 6-Oxa-3-azabicyclo[3.1.0]hexane-3-carboxylic acid tert-Butyl ester, tert-Butyl 6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate, 1-(tert-Butoxycarbonyl)-3,4-epoxypyrrolidine, 1-Boc-3,4-epoxypyrrolidine

B5385-1G B5385-5G
TCB5385-1GEA 66.36 CAD
TCB5385-1G TCB5385-5G
tert-Butyl-6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate ≥95.0% (by GC)
tert-Butyl-6-oxa-3-azabicyclo[3.1.0]hexane-3-carboxylate
Formula: C₉H₁₅NO₃
MW: 185.2203 g/mol
MDL Number: MFCD08691407
CAS Number: 114214-49-2

Order Now

Specification Test Results

Appearance Colorless to Orange to Green clear liquid
Purity (GC) min. 95.0 %
Acid Value max. 3.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR