(R)-3-Methylmorpholine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

M2847-1G M2847-200MG
76220-276EA 219.62 CAD
76220-276 76220-278
(R)-3-Methylmorpholine hydrochloride ≥98.0% (by titrimetric analysis)
(R)-3-Methylmorpholine hydrochloride
Formula: C₅H₁₂ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD18382515
CAS Number: 953780-78-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 123.0 to 127.0 deg-C
Specific rotation [a]20/D +1.5 to +3.5 deg (C=1,H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR