1-Nitropyrazole ≥97.0% (by GC)

Supplier: TCI America
Warning

N0965-25G N0965-5G
76220-364EA 507.18 CAD
76220-364 76220-366
1-Nitropyrazole ≥97.0% (by GC)
1-Nitropyrazole
Formula: C₃H₃N₃O₂
MW: 113.0748 g/mol
MDL Number: MFCD00015894
CAS Number: 7119-95-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 92.0 to 96.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR