1-Benzyl-3-methylpiperidin-4-one ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

B4465-1G B4465-5G
TCB4465-1GEA 90.06 CAD
TCB4465-1G TCB4465-5G
1-Benzyl-3-methylpiperidin-4-one ≥98.0% (by GC, titration analysis)
1-Benzyl-3-methylpiperidin-4-one
Formula: C₁₃H₁₇NO
Boiling Pt: 99°C
Storage Temperature: Refrigerator
MDL Number: MFCD00044806
CAS Number: 34737-89-8
EINECS: 252-176-5
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR