Methyl-N-boc-piperidine-3-carboxylate ≥98.0% (by GC)

Supplier: TCI America

M2918-25G M2918-5G
TCM2918-25GEA 153.26 CAD
TCM2918-25G TCM2918-5G
Methyl-N-boc-piperidine-3-carboxylate ≥98.0% (by GC)
Methyl-N-boc-piperidine-3-carboxylate
Formula: C₁₂H₂₁NO₄
MDL Number: MFCD06795926
CAS Number: 148763-41-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR