9-Bromo-10-(naphthalen-1-yl)anthracene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 9-Bromo-10-(1-naphthyl)anthracene

B4451-1G B4451-5G
TCB4451-1GEA 51.19 CAD
TCB4451-1G TCB4451-5G
9-Bromo-10-(naphthalen-1-yl)anthracene ≥98.0% (by GC)
9-Bromo-10-(naphthalen-1-yl)anthracene
Formula: C₂₄H₁₅Br
MW: 383.29 g/mol
MDL Number: MFCD11046571
CAS Number: 400607-04-7

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 177.0 to 181.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR