5-Acetonyl-2-methoxybenzene sulfonamide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
A2948-25G
A2948-5G
TCA2948-25GEA
628.84
CAD
TCA2948-25G
TCA2948-5G
5-Acetonyl-2-methoxybenzene sulfonamide ≥98.0% (by HPLC, total nitrogen)
5-Acetonyl-2-methoxybenzene sulfonamide
Formula:
C₁₀H₁₃NO₄S |
MDL Number:
MFCD07782136 CAS Number: 116091-63-5 |
Specification Test Results
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...