Ethyl-2-(2-aminothiazol-4-yl)-2-oxoacetate ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

E1159-25G E1159-5G
TCE1159-25GEA 570.38 CAD
TCE1159-25G TCE1159-5G
Ethyl-2-(2-aminothiazol-4-yl)-2-oxoacetate ≥96.0% (by HPLC, titration analysis)
Ethyl-2-(2-aminothiazol-4-yl)-2-oxoacetate
Formula: C₇H₉NO₄S
MDL Number: MFCD00010414
CAS Number: 64987-08-2

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 144.0 to 148.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR