Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America
Warning

M2531-25G M2531-5G
TCM2531-25GEA 153.26 CAD
TCM2531-25G TCM2531-5G
Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride ≥98.0% (by HPLC)
Methyl-1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
Formula: C₁₄H₁₈ClNO₃ C₁₅H₁₉NO₃·HCl
MDL Number: MFCD00012799
CAS Number: 3939-01-3
EINECS: 223-522-2

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR