Isopropylhydrazine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

I0964-1G I0964-5G
TCI0964-1GEA 33.18 CAD
TCI0964-1G TCI0964-5G
Isopropylhydrazine hydrochloride ≥98.0% (by titrimetric analysis)
Isopropylhydrazine hydrochloride
Formula: C₃H₁₁ClN₂
MDL Number: MFCD01746887
CAS Number: 16726-41-3
EINECS: 240-784-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Potassium iodate Method) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 121.0 to 125.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR