Methyl-DL-pyroglutamate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
M2720-1G
M2720-5G
TCM2720-1GEA
55.3
CAD
TCM2720-1G
TCM2720-5G
Methyl-DL-pyroglutamate ≥98.0% (by GC)
Methyl-DL-pyroglutamate
Formula:
C₆H₉NO₃ Boiling Pt: 135°C |
MDL Number:
MFCD03844651 CAS Number: 54571-66-3 EINECS: 259-233-3 |
Specification Test Results
Appearance | Colorless to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...