Methyl-1-benzylpyrrolidine-3-carboxylate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2866-1G M2866-5G
TCM2866-1GEA 112.18 CAD
TCM2866-1G TCM2866-5G
Methyl-1-benzylpyrrolidine-3-carboxylate ≥98.0% (by GC, titration analysis)
Methyl-1-benzylpyrrolidine-3-carboxylate
Formula: C₁₃H₁₇NO₂
Boiling Pt: 105°C
MDL Number: MFCD04038678
CAS Number: 17012-21-4

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR