(S)-Ethyl-N-boc-pyroglutamate ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

E1135-1G E1135-5G
TCE1135-1GEA 33.18 CAD
TCE1135-1G TCE1135-5G
(S)-Ethyl-N-boc-pyroglutamate ≥97.0% (by HPLC, total nitrogen)
(S)-Ethyl-N-boc-pyroglutamate
Formula: C₁₂H₁₉NO₅
MDL Number: MFCD07778090
CAS Number: 144978-12-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 52.0 to 56.0 deg-C
Specific rotation [a]20/D -42.0 to -46.0 deg (C=0.5、MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR