7,8-Dimethoxy-1,3-dihydro-2H-3-benzazepin-2-one ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 7,8-Dimethoxy-1H-benzo[d]azepin-2(3H)-one, 1,3-Dihydro-7,8-dimethoxy-2H-3-benzazepin-2-one

D4824-1G D4824-5G
TCD4824-1GEA 131.14 CAD
TCD4824-1G TCD4824-5G
7,8-Dimethoxy-1,3-dihydro-2H-3-benzazepin-2-one ≥98.0% (by GC)
7,8-Dimethoxy-1,3-dihydro-2H-3-benzazepin-2-one
Formula: C₁₂H₁₃NO₃
MW: 219.2400 g/mol
MDL Number: MFCD02091565
CAS Number: 73942-87-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 244.0 to 248.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR