ethyl-2-((tert-butoxycarbonyl)amino)acetate ≥98.0% (by GC)
Supplier: TCI America
B4782-25G
B4782-5G
TCB4782-25GEA
349.18
CAD
TCB4782-25G
TCB4782-5G
ethyl-2-((tert-butoxycarbonyl)amino)acetate ≥98.0% (by GC)
ethyl-2-((tert-butoxycarbonyl)amino)acetate
Formula:
C₉H₁₇NO₄ Boiling Pt: 97°C |
MDL Number:
MFCD05663720 CAS Number: 14719-37-0 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...