Boc-L-pyroglutamic acid methyl ester ≥98.0% (by GC)
Supplier: TCI America
M2803-25G
M2803-5G
TCM2803-25GEA
376.04
CAD
TCM2803-25G
TCM2803-5G
Boc-L-pyroglutamic acid methyl ester ≥98.0% (by GC)
Boc-L-pyroglutamic acid methyl ester
Formula:
C₁₁H₁₇NO₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD06809720 CAS Number: 108963-96-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 69.0 to 73.0 deg-C |
Specific rotation [a]20/D | -30.0 to -34.0 deg (C=1, CHCL3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...