1-Ethyl-1H-pyrazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1-Ethylpyrazole

E1037-25ML E1037-5ML
TCE1037-25MLEA 120.08 CAD
TCE1037-25ML TCE1037-5ML
1-Ethyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
1-Ethyl-1H-pyrazole
Formula: C₅H₈N₂
MW: 96.1304 g/mol
Boiling Pt: 137°C
MDL Number: MFCD03444367
CAS Number: 2817-71-2
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid to cloudy liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR