2,3-Dimethyl-4-fluoronitrobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

D5187-1G D5187-5G
76219-226EA 75.84 CAD
76219-226 76219-228
2,3-Dimethyl-4-fluoronitrobenzene ≥98.0% (by GC)
2,3-Dimethyl-4-fluoronitrobenzene
Formula: C₈H₈FNO₂
MW: 169.16 g/mol
Boiling Pt: 110°C
MDL Number: MFCD05663712
CAS Number: 1736-87-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR