Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine ≥98.0% (by HPLC)

Supplier: TCI America
Warning

D5036-1G D5036-5G
TCD5036-1GEA 110.6 CAD
TCD5036-1G TCD5036-5G
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine ≥98.0% (by HPLC)
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine
Formula: C₂₆H₃₅O₂P
MDL Number: MFCD05861611
CAS Number: 657408-07-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 163.0 to 167.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR