tert-Butyl-2-amino-4-fluorophenylcarbamate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: tert-Butyl (2-Amino-4-fluorophenyl)carbamate, N1-Boc-4-fluoro-1,2-phenylenediamine, tert-Butyl 2-amino-4-fluorophenylcarbamate, (2-Amino-4-fluorophenyl)carbamic acid tert-Butyl ester, N1-(tert-Butoxycarbonyl)-4-fluoro-1,2-phenylenediamine

B4236-1G B4236-200MG
TCB4236-1GEA 279.66 CAD
TCB4236-1G TCB4236-200MG
tert-Butyl-2-amino-4-fluorophenylcarbamate ≥98.0% (by titrimetric analysis)
tert-Butyl-2-amino-4-fluorophenylcarbamate
Formula: C₁₁H₁₅FN₂O₂
MW: 226.2500 g/mol
MDL Number: MFCD09701246
CAS Number: 579474-47-8

Order Now

Specification Test Results

Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 135.0 to 139.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR