6-Bromo-2-chloropyridin-3-amine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
A2967-1G
A2967-5G
TCA2967-1GEA
53.72
CAD
TCA2967-1G
TCA2967-5G
6-Bromo-2-chloropyridin-3-amine ≥98.0% (by GC)
6-Bromo-2-chloropyridin-3-amine
Formula:
C₅H₄BrClN₂ |
MDL Number:
MFCD04971930 CAS Number: 169833-70-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 117.0 to 121.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...