(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: N-(tert-Butoxycarbonyl)-D-proline methyll ester, Boc-D-Pro-OMe, N-Boc-D-proline methyll ester, N-Boc-D-proline methyl ester

B4787-1G B4787-5G
TCB4787-1GEA 42.66 CAD
TCB4787-1G TCB4787-5G
(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate ≥98.0% (by GC)
(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate
Formula: C₁₁H₁₉NO₄
MW: 229.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00190828
CAS Number: 73323-65-6

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid to cloudy liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +64.0 to +67.0 deg (C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR