Boc-L-Pro-OMe ≥97.0% (by GC)

Supplier: TCI America

B5228-25G B5228-5G
TCB5228-25GEA 148.52 CAD
TCB5228-25G TCB5228-5G
Boc-L-Pro-OMe ≥97.0% (by GC)
Boc-L-Pro-OMe
Formula: C₁₁H₁₉NO₄
Boiling Pt: 135°C
Storage Temperature: Refrigerator
MDL Number: MFCD01075092
CAS Number: 59936-29-7

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Optical purity (LC) min. 98.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR