3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M2634-1G M2634-5G
TCM2634-1GEA 58.46 CAD
TCM2634-1G TCM2634-5G
3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)aniline ≥98.0% (by GC, titration analysis)
3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)aniline
Formula: C11H10F3N3
MDL Number: MFCD11846236
CAS Number: 641571-11-1

Order Now

Specification Test Results

Appearance White to Light gray to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 129.0 to 133.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR