Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
C3243-1G
C3243-5G
76218-920EA
134.3
CAD
76218-920
76218-922
Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate ≥98.0% (by GC, titration analysis)
Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5,6-dihydropyridine-1(2H)-carboxylate
Formula:
C₁₉H₂₆BNO₄ |
MDL Number:
MFCD11521562 CAS Number: 286961-15-7 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 93.0 to 97.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...