(S)-Di-tert-butyl-5-oxopyrrolidine-1,2-dicarboxylate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B5216-1G B5216-5G
TCB5216-1GEA 69.52 CAD
TCB5216-1G TCB5216-5G
(S)-Di-tert-butyl-5-oxopyrrolidine-1,2-dicarboxylate ≥98.0% (by HPLC, total nitrogen)
(S)-Di-tert-butyl-5-oxopyrrolidine-1,2-dicarboxylate
Formula: C₁₄H₂₃NO₅
MDL Number: MFCD03094770
CAS Number: 91229-91-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 55.0 to 59.0 deg-C
Specific rotation [a]20/D -38.0 to -43.0 deg (C=1,CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR