4-Chloro-3-methoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C3304-1G C3304-5G
TCC3304-1GEA 63.2 CAD
TCC3304-1G TCC3304-5G
4-Chloro-3-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Chloro-3-methoxybenzoic acid
Formula: C₈H₇ClO₃
MDL Number: MFCD00269640
CAS Number: 85740-98-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 217.0 to 221.0 deg-C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR