3-Chloro-5-(trifluoromethyl)pyridine ≥97.0% (by GC)
Supplier: TCI America
|
Danger
|
C2951-1G
C2951-5G
TCC2951-1GEA
60.04
CAD
TCC2951-1G
TCC2951-5G
3-Chloro-5-(trifluoromethyl)pyridine ≥97.0% (by GC)
3-Chloro-5-(trifluoromethyl)pyridine
Formula:
C6H3ClF3N Flash Pt: 54°C |
MDL Number:
MFCD00042227 CAS Number: 85148-26-1 UN: 1992 ADR: 3,III |
Specification Test Results
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...