3-Fluoroazetidine hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

Synonyms: 3-Fluoroazetidine HCl

F1186-1G F1186-200MG
TCF1186-1GEA 216.46 CAD
TCF1186-1G TCF1186-200MG
3-Fluoroazetidine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
3-Fluoroazetidine hydrochloride
Formula: C3H7ClFN
MW: 111.55 g/mol
MDL Number: MFCD05663715
CAS Number: 617718-46-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 128.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR