2-(3-Bromophenyl)pyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

B5113-1G B5113-5G
76218-822EA 94.8 CAD
76218-822 76218-824
2-(3-Bromophenyl)pyridine ≥98.0% (by GC)
2-(3-Bromophenyl)pyridine
Formula: C₁₁H₈BrN
MDL Number: MFCD09749846
CAS Number: 4373-60-8

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR