2-(Piperidin-4-yl)-1H-benzo[d]imidazole ≥98.0% (by GC)

Supplier: TCI America

P2382-1G P2382-5G
76220-416EA 186.44 CAD
76220-416 76220-418
2-(Piperidin-4-yl)-1H-benzo[d]imidazole ≥98.0% (by GC)
2-(Piperidin-4-yl)-1H-benzo[d]imidazole
Formula: C₁₂H₁₅N₃
MDL Number: MFCD01475843
CAS Number: 38385-95-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR