1,2,3,4-Tetrahydroquinoxaline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

T2999-1G T2999-5G
TCT2999-1GEA 112.34 CAD
TCT2999-1G TCT2999-5G
1,2,3,4-Tetrahydroquinoxaline ≥98.0% (by GC, titration analysis)
1,2,3,4-Tetrahydroquinoxaline
Formula: C₈H₁₀N₂
Boiling Pt: 154°C
MDL Number: MFCD00047564
CAS Number: 3476-89-9

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 95.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR